Learn More
Medchemexpress LLC Estradiol benzoate-d3 | 1192354-74-7 | 99.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB1192S1MG
Encompass_Preferred
Estradiol benzoate-d3 is the deuterium labeled Estradiol benzoate. It is a proagent of estradiol that acts as a steroid sex hormone. This compound exhibits mild anabolic and metabolic properties, and increases blood coagulability. It is for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-13991