Learn More
Medchemexpress LLC Estradiol benzoate-d3 | 1192354-74-7 | 99.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1192S5MG
Encompass_Preferred
Estradiol benzoate-d3 is the deuterium labeled Estradiol benzoate. Estradiol Benzoate (β-Estradiol 3-benzoate), a proagent of estradiol, acts as a steroid sex hormone. It exhibits mild anabolic and metabolic properties, and increases blood coagulability.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-12423