Learn More
Medchemexpress LLC Estradiol benzoate (standard) | 50-50-0 | 99.7% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1192R100MG
Encompass_Preferred
Estradiol benzoate (Standard) is the analytical standard of Estradiol benzoate, intended for research and analytical applications. Estradiol benzoate is a HBx protein inhibitor and inhibits androgen and hepatitis B virus (HBV) transcription and replication. It also exhibits antifertility effects, anti-*Toxoplasma gondii* activity, and can improve memory behavior in Ovariectomy (Ovx) female mice. It is for research use only.
  • Analytical standard for Estradiol benzoate.
  • HBx protein inhibitor.
  • Inhibits androgen and hepatitis B virus (HBV) transcription and replication.
  • Shows antifertility effects.
  • Exhibits anti-*Toxoplasma gondii* activity.
  • Can improve memory behavior of Ovariectomy (Ovx) female mice.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-14160