Learn More
Medchemexpress LLC Estradiol benzoate (standard) | 50-50-0 | 99.7% | 50 MG
Supplier: Medchemexpress LLC HYB1192R50MG
Estradiol benzoate (Standard) is an analytical standard intended for research and analytical applications. It acts as an HBx protein inhibitor, inhibiting androgen and hepatitis B virus (HBV) transcription and replication. Additionally, it demonstrates antifertility effects, anti-Toxoplasma gondii activity, and can enhance memory behavior in Ovariectomy (Ovx) female mice. This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.
- Intended for research and analytical applications.
- Inhibits HBx protein, androgen, and hepatitis B virus (HBV) transcription and replication.
- Demonstrates antifertility effects.
- Shows anti-Toxoplasma gondii activity.
- Improves memory behavior in Ovariectomy (Ovx) female mice.
- Suitable for HPLC, GC, and MS research experiments.