Learn More
Cayman Chemical Estradiol Cypionate, Purity: ≥98%, CAS: 313-06-4, 1 g
SDP
Catalog No. 502978789
Encompass_Preferred
A synthetic estrogen; reduces estrogen-depletion induced spikes in basal tail temperature in ovariectomized wild-type, ERα-/-, and ERβ-/- mice at 0.1 mg/kg per day; increases the rate of pregnancy in cows induced to ovulate small dominant follicles; reduces edema and articular joint neutrophil infiltration in an ovariectomized mouse model of zymosan-induced arthritis
Catalog No. 50-297-8789 Supplier Cayman Chemical Supplier No. 311051 G
A synthetic estrogen; reduces estrogen-depletion induced spikes in basal tail temperature in ovariectomized wild-type, ERα-/-, and ERβ-/- mice at 0.1 mg/kg per day; increases the rate of pregnancy in cows induced to ovulate small dominant follicles; reduces edema and articular joint neutrophil infiltration in an ovariectomized mouse model of zymosan-induced arthritis