Learn More
Apexbio Technology LLC Estradiol Cypionate 313-06-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000087656
Encompass_Preferred
Estradiol cypionate (313-06-4) is a small-molecule agonist targeting estrogen receptors (ER) It is designed to activate ER thereby modulating downstream ER-mediated gene transcription and signaling pathways and regulating processes such as vascular homeostasis Estradiol cypionate exerts its biological activity primarily through ER agonism influencing gene expression and inhibiting endothelin-1 (ET-1) synthesis via ER-dependent mechanisms Based on these pharmacological properties estradiol cypionate holds research potential in elucidating estrogen receptor mechanisms and studying estrogen-associated vascular and endocrine systems and disorders
Catalog No. 50-000-87656 Supplier Apexbio Technology LLC Supplier No. B15015.1
Estradiol cypionate (313-06-4) is a small-molecule agonist targeting estrogen receptors (ER) It is designed to activate ER thereby modulating downstream ER-mediated gene transcription and signaling pathways and regulating processes such as vascular homeostasis Estradiol cypionate exerts its biological activity primarily through ER agonism influencing gene expression and inhibiting endothelin-1 (ET-1) synthesis via ER-dependent mechanisms Based on these pharmacological properties estradiol cypionate holds research potential in elucidating estrogen receptor mechanisms and studying estrogen-associated vascular and endocrine systems and disorders