Learn More
Medchemexpress LLC Estradiol cypionate | 313-06-4 | MFCD00056558 | 99.5% | 1 ML
SDP
Catalog No. 5000413345
Encompass_Preferred
Estradiol cypionate is the 17β-cypionate ester of Estradiol, inhibiting ET-1 synthesis by acting on estrogen receptors. It is intended for research use only. In vitro, it inhibits proliferation of gastric cancer cells and induces G1/S phase cell cycle arrest and apoptosis by affecting the PI3K/Akt/mTOR pathway. In vivo, it increases cortical bone density in ovariectomized rabbits, increases occurrence of estrus and pregnancy in cows, and inhibits tumor growth in MGC803 nude mice tumor models.
  • Inhibits gastric cancer cell proliferation
  • Induces G1/S phase cell cycle arrest and apoptosis
  • Increases cortical bone density
  • Promotes estrus and pregnancy
  • Inhibits tumor growth
Catalog No. 50-004-13345 Supplier Medchemexpress LLC Supplier No. HYB110010MM/1ML
Estradiol cypionate is the 17β-cypionate ester of Estradiol, inhibiting ET-1 synthesis by acting on estrogen receptors. It is intended for research use only. In vitro, it inhibits proliferation of gastric cancer cells and induces G1/S phase cell cycle arrest and apoptosis by affecting the PI3K/Akt/mTOR pathway. In vivo, it increases cortical bone density in ovariectomized rabbits, increases occurrence of estrus and pregnancy in cows, and inhibits tumor growth in MGC803 nude mice tumor models.
  • Inhibits gastric cancer cell proliferation
  • Induces G1/S phase cell cycle arrest and apoptosis
  • Increases cortical bone density
  • Promotes estrus and pregnancy
  • Inhibits tumor growth