Learn More
Cayman Chemical EstradIol CypIonate 500mg
SDP
Catalog No. 502978809
Encompass_Preferred
A synthetic estrogen; reduces estrogen-depletion induced spikes in basal tail temperature in ovariectomized wild-type, ERα-/-, and ERβ-/- mice at 0.1 mg/kg per day; increases the rate of pregnancy in cows induced to ovulate small dominant follicles; reduces edema and articular joint neutrophil infiltration in an ovariectomized mouse model of zymosan-induced arthritis
Catalog No. 50-297-8809 Supplier Cayman Chemical Supplier No. 31105500 MG
A synthetic estrogen; reduces estrogen-depletion induced spikes in basal tail temperature in ovariectomized wild-type, ERα-/-, and ERβ-/- mice at 0.1 mg/kg per day; increases the rate of pregnancy in cows induced to ovulate small dominant follicles; reduces edema and articular joint neutrophil infiltration in an ovariectomized mouse model of zymosan-induced arthritis