Learn More
Selleck Chemical LLC Estradiol Cypionate S4046-1g
SDP
Catalog No. 503109473
Encompass_Preferred
Estradiol cypionate is the 17 -cyclopentylpropinate ester of estradiol which inhibits ET-1 synthesis via estrogen receptor
Catalog No. 50-310-9473 Supplier Selleck Chemical LLC Supplier No. S40461G
Add to Cart
Add to Cart
Estradiol cypionate is the 17 -cyclopentylpropinate ester of estradiol which inhibits ET-1 synthesis via estrogen receptor