Learn More
Medchemexpress LLC Estradiol-d2-1 | 3188-46-3 | 1 MG
Supplier: Medchemexpress LLC HYB0141S61MG
Estradiol-d2-1 is a deuterium-labeled Estradiol, a steroid sex hormone vital to the maintenance of fertility and secondary sexual characteristics in females. It upregulates IL-6 expression through the estrogen receptor β (ERβ) pathway.
- Can be used as a tracer
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
- Incorporates stable heavy isotopes into drug molecules
- Has potential to affect pharmacokinetic and metabolic profiles of drugs