Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estradiol-d3 | 79037-37-9 | 98.6% | 5 MG
SDP

Catalog No. 5000450697
Encompass_Preferred

Estradiol-d3 is a deuterium-labeled form of Estradiol, a steroid sex hormone crucial for maintaining fertility and secondary sexual characteristics in females. It upregulates IL-6 expression via the estrogen receptor β (ERβ) pathway and is intended for research use only. This compound serves as a tracer and internal standard for quantitative analysis, leveraging the potential of deuteration to influence pharmacokinetic and metabolic profiles.

  • Used as a tracer
  • Used as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Deuteration influences drug pharmacokinetic and metabolic profiles
  • Appears as a white to off-white solid
  • Molecular weight: 275.40
  • Chemical formula: C18H21D3O2
  • Powder storage: -20°C for 3 years or 4°C for 2 years
  • Solvent storage: -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO (50 mg/mL) and ethanol (20 mg/mL)

Catalog No. 50-004-50697 Supplier Medchemexpress LLC Supplier No. HYB0141S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Estradiol-d3 is a deuterium-labeled form of Estradiol, a steroid sex hormone crucial for maintaining fertility and secondary sexual characteristics in females. It upregulates IL-6 expression via the estrogen receptor β (ERβ) pathway and is intended for research use only. This compound serves as a tracer and internal standard for quantitative analysis, leveraging the potential of deuteration to influence pharmacokinetic and metabolic profiles.

  • Used as a tracer
  • Used as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Deuteration influences drug pharmacokinetic and metabolic profiles
  • Appears as a white to off-white solid
  • Molecular weight: 275.40
  • Chemical formula: C18H21D3O2
  • Powder storage: -20°C for 3 years or 4°C for 2 years
  • Solvent storage: -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO (50 mg/mL) and ethanol (20 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.