Learn More
Medchemexpress LLC Estradiol-d4 | 66789-03-5 | 99.6% | 1 MG
SDP
Catalog No. 5000450474
Encompass_Preferred
Estradiol-d4 is the deuterium labeled Estradiol. Estradiol is a steroid sex hormone vital to the maintenance of fertility and secondary sexual characteristics in females. It upregulates IL-6 expression through the estrogen receptor beta (ERβ) pathway.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements
  • Deuteration affects the pharmacokinetic and metabolic profiles of drugs
  • Applicable in cancer research, metabolic or endocrine disease, and endocrinology
Catalog No. 50-004-50474 Supplier Medchemexpress LLC Supplier No. HYB0141S11MG
Estradiol-d4 is the deuterium labeled Estradiol. Estradiol is a steroid sex hormone vital to the maintenance of fertility and secondary sexual characteristics in females. It upregulates IL-6 expression through the estrogen receptor beta (ERβ) pathway.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements
  • Deuteration affects the pharmacokinetic and metabolic profiles of drugs
  • Applicable in cancer research, metabolic or endocrine disease, and endocrinology