Learn More
Medchemexpress LLC Estradiol-d5 | 221093-45-4 | 99.1% | 1 MG
SDP
Catalog No. 5000450471
Encompass_Preferred
Estradiol-d5 is a deuterium-labeled form of Estradiol. Estradiol is a steroid sex hormone crucial for maintaining fertility and secondary sexual characteristics in females. It upregulates IL-6 expression through the estrogen receptor β (ERβ) pathway.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for quantitation during the drug development process.
Catalog No. 50-004-50471 Supplier Medchemexpress LLC Supplier No. HYB0141S21MG
Estradiol-d5 is a deuterium-labeled form of Estradiol. Estradiol is a steroid sex hormone crucial for maintaining fertility and secondary sexual characteristics in females. It upregulates IL-6 expression through the estrogen receptor β (ERβ) pathway.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for quantitation during the drug development process.