Learn More
Medchemexpress LLC Estradiol-d5 | 221093-45-4 | >99.7% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0141S25MG
Encompass_Preferred
Estradiol-d5 is deuterium labeled Estradiol. Estradiol is a steroid sex hormone crucial for maintaining fertility and secondary sexual characteristics in females. It upregulates IL-6 expression through the estrogen receptor β (ERβ) pathway. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound.
Catalog No. 50-004-50910