Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estradiol dipropionate | 113-38-2 | 99.5% | 100 MG
SDP

Catalog No. 5000409723
Encompass_Preferred

Estradiol dipropionate is a combined estrogen-progesterone, acting as an estrogen and progesterone agonist. It is intended for research use only.

  • Purity: 99.5%
  • Molecular weight: 384.51
  • Formula: C24H32O4
  • Appearance: White to off-white solid
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage for in solvent: -80°C for 2 years, -20°C for 1 year
  • Shipping at room temperature is acceptable if less than 2 weeks

Catalog No. 50-004-09723 Supplier Medchemexpress LLC Supplier No. HYB2245100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Estradiol dipropionate is a combined estrogen-progesterone, acting as an estrogen and progesterone agonist. It is intended for research use only.

  • Purity: 99.5%
  • Molecular weight: 384.51
  • Formula: C24H32O4
  • Appearance: White to off-white solid
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage for in solvent: -80°C for 2 years, -20°C for 1 year
  • Shipping at room temperature is acceptable if less than 2 weeks

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.