Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedical™ Estradiol (E2) Coated Tube RIA Kit

Catalog No. MP07238105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 Tubes
500 Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP07238105 500 Tubes
MP07238102 100 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP07238105 Supplier MP Biomedical™ Supplier No. 07238105
Only null left
Add to Cart
Add to Cart

Quantitative Solid Phase Radioimmunoassay

The Estradiol Coated Tube RIA Kit is intended for the measurement of the unconjugated form of E2 (17β-Estradiol) in a sample. In the ovulating woman, circulating E2 originates predominantly from cohorts of maturing follicles and ultimately from a single follicle recruited for ovulation and the corpus luteum it forms. E2 concentrations are highly variable depending on the phase of the menstrual cycle. In the postmenopausal woman, E2 originates from extraglandular conversion of androgens and circulates in low, noncyclic concentrations. In prepubertal children and in the male, circulating E2 concentrations are low and non-cyclic.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Kit and Assay
Sample Type Plasma, Serum
Target Species Human
For Use With (Application) Endocrinology
Quantity 500 Tubes
Sample Volume 100 μL
Storage Requirements 2°C to 8°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.