Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Estradiol E2 ELISA Kit

Catalog No. KAQ0622
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KAQ0622 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KAQ0622 Supplier Invitrogen™ Supplier No. KAQ0622
Add to Cart
Edge
Add to Cart

ELISA

The Human Estradiol E2 ELISA Kit is a solid-phase sandwich Enzyme-Linked Immunosorbent Assay (ELISA). This assay is designed to detect and quantify the level of 17-beta-estradiol (E2) in serum and plasma. The assay will recognize both natural and recombinant human estradiol E2. Principle of the method The Human Estradiol solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Estradiol (17 beta-estradiol) is a sex hormone. Estradiol is the predominant sex hormone present in females. It is also present in males, being produced as an active metabolic product of testosterone. It represents the major estrogen in humans. Estradiol has not only a critical impact on reproductive and sexual functioning, but also affects other organs including the bones.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 25-2000 pg/mL
Assay Sensitivity 10.6 pg/mL
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Hormones & Receptors
Sample Type Plasma, Serum
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 17-Beta-Estradiol; 17-beta-HSD17-Beta-Estradiol; 17-beta-HSD;
Interassay CV 8.9%
Intraassay CV 11%
Kit Contents Pre-coated 96 well strip-well plate, Standard, Controls, SAV-HRP, Wash Buffer, Chromogen, Stop Solution
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Shipping Condition Wet Ice
Storage Requirements 2-8°C
Target Estradiol
Test Time 1 hr 20 min
Total Assay Time 2 hr
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.