Learn More
Arbor Assays Estradiol Serum ELISA Kit (1 Plate)
SDP
Supplier:  Arbor Assays KB30H1
Encompass_Preferred
The DetectX Serum 17β-Estradiol ELISA Kits quantitatively measure free 17β-Estradiol in serum and plasma.
Catalog No. 50-227-8531