Learn More
Drg International Inc DRG Estradiol ELISA, 96 Well Kit
Supplier:  Drg International Inc EIA2693
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of estradiol in serum and plasma. The DRG Estradiol ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a polyclonal [rabbit] antibody directed towards an antigenic site of the estradiol molecule.
Catalog No. 50-187-1233