Learn More
Cayman Chemical Estradiol ELISA Kit, 96 solid
SDP
Catalog No. 502896799
Encompass_Preferred
Cayman’s Estradiol ELISA Kit is a competitive assay that can be used for the quantification of estradiol in plasma and serum. This assay has a range of 0.61-10,000 pg/ml with a sensitivity of approximately 20 pg/ml (80% B/B0)
Catalog No. 50-289-6799 Supplier Cayman Chemical Supplier No. 50189096 SOLID
Cayman’s Estradiol ELISA Kit is a competitive assay that can be used for the quantification of estradiol in plasma and serum. This assay has a range of 0.61-10,000 pg/ml with a sensitivity of approximately 20 pg/ml (80% B/B0)