Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estradiol enanthate (E2EN) | 4956-37-0 | 98.1% | 500 MG
SDP

Catalog No. 5000412154
Encompass_Preferred

Estradiol enanthate (E2EN) is a shorter-acting estrogen. It is an injectable contraceptive when combined with Dihydroxyprogesterone acetophenide. It promotes withdrawal bleeding similar to menstruation and is intended for research use only.

  • Molecular weight: 384.55
  • Formula: C25H36O3
  • Appearance: Solid
  • Color: White to off-white
  • Solubility in DMSO: 125 mg/mL (325.06 mM; requires ultrasonic)
  • Solubility in ethanol: ≥ 100 mg/mL (260.04 mM)
  • Shipping: Room temperature in continental US
  • Storage: 4°C, sealed storage, away from moisture and light. In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture and light).

Catalog No. 50-004-12154 Supplier Medchemexpress LLC Supplier No. HYB1830500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Estradiol enanthate (E2EN) is a shorter-acting estrogen. It is an injectable contraceptive when combined with Dihydroxyprogesterone acetophenide. It promotes withdrawal bleeding similar to menstruation and is intended for research use only.

  • Molecular weight: 384.55
  • Formula: C25H36O3
  • Appearance: Solid
  • Color: White to off-white
  • Solubility in DMSO: 125 mg/mL (325.06 mM; requires ultrasonic)
  • Solubility in ethanol: ≥ 100 mg/mL (260.04 mM)
  • Shipping: Room temperature in continental US
  • Storage: 4°C, sealed storage, away from moisture and light. In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture and light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.