Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Estradiol, MP Biomedicals™

Catalog No. ICN10165680 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10165680 100 g
ICN10165605 5 g
ICN10165625 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10165680 Supplier MP Biomedicals, Inc Supplier No. 0210165680
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50-28-2
Molecular Formula C18H24O2
Molecular Weight (g/mol) 272.388
InChI Key VOXZDWNPVJITMN-ZBRFXRBCSA-N
Synonym estradiol, beta-estradiol, 17beta-estradiol, oestradiol, dihydrofolliculin, estrace, ovocyclin, progynon, dihydrotheelin, dihydroxyestrin
PubChem CID 5757
ChEBI CHEBI:16469
IUPAC Name (8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
SMILES CC12CCC3C(C1CCC2O)CCC4=C3C=CC(=C4)O

Specifications

Quantity 100 g
Formula Weight 272.4
Physical Form Solid
Chemical Name or Material β-Estradiol
storageNote1 Store under nitrogen.
Recommended Storage Store at room temperature (15° to 30°C).
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.