Learn More
Alpco Ltd Estradiol Rat ELISA
SDP
Supplier:  Alpco Ltd 55ESTRTE01
Encompass_Preferred
The Estradiol Rat ELISA is an Enzyme Immunoassay for the quantitative determination of estradiol in rat serum.
  • Includes standards
  • Features an analytical sensitivity of 2.5 pg/mL
  • Broad detection range of 2.5 – 1280 pg/mL
  • No extraction of samples needed
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2318921