Learn More
Medchemexpress LLC Estradiol (Standard) | 50-28-2 | 100.0% | 50 MG
SDP
Catalog No. 5000450929
Encompass_Preferred
Estradiol (Standard) is the analytical standard of Estradiol, intended for research and analytical applications. As a steroid hormone and major female sex hormone, it can up-regulate the expression of neural markers of human endometrial stem cells (hEnSCs) and promote their neural differentiation. It is suitable for the research of cancers, neurodegenerative diseases, and neural tissue engineering.
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments
  • Applications include HPLC, GC and MS
Catalog No. 50-004-50929 Supplier Medchemexpress LLC Supplier No. HYB0141R50MG
Add to Cart
Add to Cart
Estradiol (Standard) is the analytical standard of Estradiol, intended for research and analytical applications. As a steroid hormone and major female sex hormone, it can up-regulate the expression of neural markers of human endometrial stem cells (hEnSCs) and promote their neural differentiation. It is suitable for the research of cancers, neurodegenerative diseases, and neural tissue engineering.
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments
  • Applications include HPLC, GC and MS