Learn More
Medchemexpress LLC Estradiol (Standard) | 50-28-2 | 99.9% | 1 ML
SDP
Catalog No. 5000450927
Encompass_Preferred
Estradiol (β-Estradiol) is a steroid hormone and the primary female sex hormone. It can increase the expression of neural markers in human endometrial stem cells (hEnSCs) and promote their neural differentiation. This product is intended for research use only and is utilized in studies concerning cancers, neurodegenerative diseases, and neural tissue engineering.
  • Increases expression of neural markers in human endometrial stem cells.
  • Promotes neural differentiation of human endometrial stem cells.
  • Utilized in research for cancers, neurodegenerative diseases, and neural tissue engineering.
  • Increases SIRT1 protein levels in C2C12 cells.
  • Increases FoxL2 and pFoxL2 protein expression levels in ZF4 cells.
  • Promotes proliferation of sheep myoblasts.
  • Enhances expression of ERRα and VLDL-related genes in primary hepatocytes.
  • Induces mRNA expression of pS2 and NRF1 genes in MCF-7 cells.
Catalog No. 50-004-50927 Supplier Medchemexpress LLC Supplier No. HYB014110MM/1ML
Estradiol (β-Estradiol) is a steroid hormone and the primary female sex hormone. It can increase the expression of neural markers in human endometrial stem cells (hEnSCs) and promote their neural differentiation. This product is intended for research use only and is utilized in studies concerning cancers, neurodegenerative diseases, and neural tissue engineering.
  • Increases expression of neural markers in human endometrial stem cells.
  • Promotes neural differentiation of human endometrial stem cells.
  • Utilized in research for cancers, neurodegenerative diseases, and neural tissue engineering.
  • Increases SIRT1 protein levels in C2C12 cells.
  • Increases FoxL2 and pFoxL2 protein expression levels in ZF4 cells.
  • Promotes proliferation of sheep myoblasts.
  • Enhances expression of ERRα and VLDL-related genes in primary hepatocytes.
  • Induces mRNA expression of pS2 and NRF1 genes in MCF-7 cells.