Learn More
Medchemexpress LLC Estradiol (Standard) | 50-28-2 | MFCD00003693 | 100.0% | 25 MG
SDP
Catalog No. 5000406518
Encompass_Preferred
Estradiol (Standard) is an analytical standard of Estradiol, a major female sex hormone and steroid hormone. Intended for research and analytical applications, it can up-regulate the expression of neural markers of human endometrial stem cells (hEnSCs) and promote their neural differentiation. This compound is useful for research in cancers, neurodegenerative diseases, and neural tissue engineering.
  • Analytical standard grade
  • Reference standard for assays
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Applicable in techniques such as HPLC, GC, and MS
  • Supports research in cancers, neurodegenerative diseases, and neural tissue engineering
  • Up-regulates neural markers of human endometrial stem cells
  • Promotes neural differentiation
Catalog No. 50-004-06518 Supplier Medchemexpress LLC Supplier No. 25MGHYB0141R
Estradiol (Standard) is an analytical standard of Estradiol, a major female sex hormone and steroid hormone. Intended for research and analytical applications, it can up-regulate the expression of neural markers of human endometrial stem cells (hEnSCs) and promote their neural differentiation. This compound is useful for research in cancers, neurodegenerative diseases, and neural tissue engineering.
  • Analytical standard grade
  • Reference standard for assays
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Applicable in techniques such as HPLC, GC, and MS
  • Supports research in cancers, neurodegenerative diseases, and neural tissue engineering
  • Up-regulates neural markers of human endometrial stem cells
  • Promotes neural differentiation