Learn More
Description
Estradiol USP comes as white or creamy white, small crystals or crystalline powder.It is odorless and hygroscopic. Estradiol USP is practically insoluble in water, is soluble in alcohol and in solutions of fixed alkali hydroxides, and finally is sparingly soluble in vegetable oils. Micronization level: 95% < 10 μm.
Alternative name(s): Estradiol hemihydrate, β-Estradiol hemihydrate, beta-Estradiol semihydrate
Specifications
Specifications
| Chemical Name or Material | Estradiol, USP (Hemihydrate) (Micronized) |
| Packaging | Bottle, Amber Glass |
| CAS | 35380-71-3 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 281.39 |
| Quantity | 1 g |
| Percent Purity | 97 to 103 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
