Learn More
Apexbio Technology LLC Estradiol valerate 979-32-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063711
Encompass_Preferred
Estradiol valerate is a synthetic estrogen ester and agonist of estrogen receptors (ERs) As a prodrug following administration it undergoes enzymatic hydrolysis to release active estradiol thereby activating estrogen receptor-mediated signaling pathways Estradiol interactions with ER and ER receptors lead to modulation of gene transcription related to cellular growth differentiation reproductive function and endocrine regulation In biomedical research Estradiol valerate frequently serves as a representative estrogenic agent to study ER-mediated biological mechanisms hormone-dependent cellular responses and experimental hormone deficiency models such as menopausal or ovarian dysfunction contexts Common research applications include investigating cellular responses to estrogen stimulation examining reproductive biology mechanisms and evaluating therapeutic strategies targeting hormone receptors
Catalog No. 50-000-63711 Supplier Apexbio Technology LLC Supplier No. B15065.1
Estradiol valerate is a synthetic estrogen ester and agonist of estrogen receptors (ERs) As a prodrug following administration it undergoes enzymatic hydrolysis to release active estradiol thereby activating estrogen receptor-mediated signaling pathways Estradiol interactions with ER and ER receptors lead to modulation of gene transcription related to cellular growth differentiation reproductive function and endocrine regulation In biomedical research Estradiol valerate frequently serves as a representative estrogenic agent to study ER-mediated biological mechanisms hormone-dependent cellular responses and experimental hormone deficiency models such as menopausal or ovarian dysfunction contexts Common research applications include investigating cellular responses to estrogen stimulation examining reproductive biology mechanisms and evaluating therapeutic strategies targeting hormone receptors