Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estradiol valerate | 979-32-8 | >99.9% | 25 MG
SDP

Catalog No. 5000450490
Encompass_Preferred

Estradiol valerate is a synthetic estrogen widely used in combination with other steroid hormones in hormone replacement therapy agents. It is for research use only and not sold to patients. It is also identified as a reproductive toxicant and estrogenic chemical in both male and female fish.

  • Molecular weight: 356.50
  • Formula: C23H32O3
  • Appearance: Solid, white to off-white
  • Shipping: Room temperature in continental US
  • Storage: Powder at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 2 years, -20°C for 1 year
  • Solubility in DMSO: ≥ 50 mg/mL (140.25 mM)
  • Solubility in ethanol: 12.5 mg/mL (35.06 mM)
  • Insoluble in H2O
  • Target: Estrogen Receptor/ERR

Catalog No. 50-004-50490 Supplier Medchemexpress LLC Supplier No. 25MGHYB0672R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Estradiol valerate is a synthetic estrogen widely used in combination with other steroid hormones in hormone replacement therapy agents. It is for research use only and not sold to patients. It is also identified as a reproductive toxicant and estrogenic chemical in both male and female fish.

  • Molecular weight: 356.50
  • Formula: C23H32O3
  • Appearance: Solid, white to off-white
  • Shipping: Room temperature in continental US
  • Storage: Powder at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 2 years, -20°C for 1 year
  • Solubility in DMSO: ≥ 50 mg/mL (140.25 mM)
  • Solubility in ethanol: 12.5 mg/mL (35.06 mM)
  • Insoluble in H2O
  • Target: Estrogen Receptor/ERR

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.