Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estradiol valerate | 979-32-8 | MFCD00056541 | 99.91% | 1 G
SDP

Catalog No. 5000409179
Encompass_Preferred

Estradiol valerate (β-Estradiol 17-valerate) is a synthetic estrogen used in hormone replacement therapy. It is also identified as a reproductive toxicant and estrogenic chemical in both male and female fish.

  • Synthetic estrogen
  • Targeted to estrogen receptor/ERR
  • Appearance as a solid, white to off-white powder
  • Solubility in DMSO: ≥ 50 mg/mL
  • Solubility in ethanol: 12.5 mg/mL (needs ultrasonic)
  • Powder storage conditions: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage conditions: -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-09179 Supplier Medchemexpress LLC Supplier No. HYB06721G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Estradiol valerate (β-Estradiol 17-valerate) is a synthetic estrogen used in hormone replacement therapy. It is also identified as a reproductive toxicant and estrogenic chemical in both male and female fish.

  • Synthetic estrogen
  • Targeted to estrogen receptor/ERR
  • Appearance as a solid, white to off-white powder
  • Solubility in DMSO: ≥ 50 mg/mL
  • Solubility in ethanol: 12.5 mg/mL (needs ultrasonic)
  • Powder storage conditions: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage conditions: -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.