Learn More
Medchemexpress LLC Estradiol valerate (standard) | 979-32-8 | 99.8% | 100 MG
SDP
Catalog No. 5000408260
Encompass_Preferred
Estradiol valerate (Standard) is the analytical standard of Estradiol valerianate. This product is intended for research and analytical applications. Estradiol valerate (β-Estradiol 17-valerate) is a synthetic estrogen widely used in combination with other steroid hormones in hormone replacement therapy agents.
  • It is the analytical standard of Estradiol valerianate.
  • Intended for research and analytical applications.
  • Used as a synthetic estrogen.
  • Widely used in combination with other steroid hormones in hormone replacement therapy.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-004-08260 Supplier Medchemexpress LLC Supplier No. HYB0672R100MG
Estradiol valerate (Standard) is the analytical standard of Estradiol valerianate. This product is intended for research and analytical applications. Estradiol valerate (β-Estradiol 17-valerate) is a synthetic estrogen widely used in combination with other steroid hormones in hormone replacement therapy agents.
  • It is the analytical standard of Estradiol valerianate.
  • Intended for research and analytical applications.
  • Used as a synthetic estrogen.
  • Widely used in combination with other steroid hormones in hormone replacement therapy.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.