Learn More
Selleck Chemical LLC Estramustine E4897-100MG
SDP
Catalog No. 5000043264
Encompass_Preferred
Estramustine(NSC-89201 LEO275) is used to treat hormone-resistant prostate cancer and stabilize microtubule dynamics It acts as an anti-mitotic and anti-microtubule agent and disrupts microtubule disassembly It inhibits the binding of MAPs 1 and 2 to taxol-stabilized microtubules hinders microtubule assembly and promotes the disassembly of MAP-2-associated microtubules
Catalog No. 50-000-43264 Supplier Selleck Chemical LLC Supplier No. E4897100MG
Estramustine(NSC-89201 LEO275) is used to treat hormone-resistant prostate cancer and stabilize microtubule dynamics It acts as an anti-mitotic and anti-microtubule agent and disrupts microtubule disassembly It inhibits the binding of MAPs 1 and 2 to taxol-stabilized microtubules hinders microtubule assembly and promotes the disassembly of MAP-2-associated microtubules