Learn More
TARGETMOL CHEMICALS INC Estramustine phosphate sodium
SDP
Catalog No. 502879027
Encompass_Preferred
Also available in 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Estramustine phosphate sodium (Ro 21-8837/001) is an antimicrotubule chemotherapy agent; arrests prostate cancer cells in the G2/M phase of the cell cycle. Purity 99.54%
Catalog No. 50-287-9027 Supplier TARGETMOL CHEMICALS INC Supplier No. T4451100MG
Also available in 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Estramustine phosphate sodium (Ro 21-8837/001) is an antimicrotubule chemotherapy agent; arrests prostate cancer cells in the G2/M phase of the cell cycle. Purity 99.54%