Learn More
Selleck Chemical LLC Estramustine phosphate sodium S2441-1g
SDP
Catalog No. 503112970
Encompass_Preferred
Estramustine phosphate sodium (EMP Emcyt Estracyt) is an antitumour drug with a unique dual mode of action Estrone and estradiol products of the metabolism of estramustine phosphate sodium have shown antigonadotrophic activity resulting in reduced testosterone levels Estramustine(the cytotoxic metabolite produced by dephosphorylation of estramustine phosphate sodium) is an antiprostatic tumor drug with antimicrotubule effects
Catalog No. 50-311-2970 Supplier Selleck Chemical LLC Supplier No. S24411G
Estramustine phosphate sodium (EMP Emcyt Estracyt) is an antitumour drug with a unique dual mode of action Estrone and estradiol products of the metabolism of estramustine phosphate sodium have shown antigonadotrophic activity resulting in reduced testosterone levels Estramustine(the cytotoxic metabolite produced by dephosphorylation of estramustine phosphate sodium) is an antiprostatic tumor drug with antimicrotubule effects