Learn More
Medchemexpress LLC Estramustine phosphate sodium | 52205-73-9 | 99.6% | 564.35 | 25 MG
SDP
Catalog No. 5000351589
Encompass_Preferred
Estramustine phosphate sodium is an estradiol analog and an orally active antimicrotubule chemotherapy agent. It works by depolymerizing microtubules through binding to microtubule-associated proteins (MAPs) and/or tubulin. This action can interfere with mitosis, trigger cell death, and induce apoptosis, making it suitable for research applications in various cancers, including prostate cancer.
  • Orally active antimicrotubule chemotherapy agent
  • Depolymerizes microtubules by binding to MAPs and/or tubulin
  • Interferes with mitosis and induces apoptosis
  • Suitable for cancer research, particularly prostate cancer
  • Store at 4°C, sealed, away from moisture and light (solid)
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-51589 Supplier Medchemexpress LLC Supplier No. 25MGHY13627
Estramustine phosphate sodium is an estradiol analog and an orally active antimicrotubule chemotherapy agent. It works by depolymerizing microtubules through binding to microtubule-associated proteins (MAPs) and/or tubulin. This action can interfere with mitosis, trigger cell death, and induce apoptosis, making it suitable for research applications in various cancers, including prostate cancer.
  • Orally active antimicrotubule chemotherapy agent
  • Depolymerizes microtubules by binding to MAPs and/or tubulin
  • Interferes with mitosis and induces apoptosis
  • Suitable for cancer research, particularly prostate cancer
  • Store at 4°C, sealed, away from moisture and light (solid)
  • In solvent, store at -80°C for 6 months or -20°C for 1 month