Learn More
Medchemexpress LLC Estramustine phosphate sodium | 52205-73-9 | MFCD00866320 | 99.4% | 564.35 g/mol | C23H30Cl2NNa2O6P | 5 MG
SDP
Catalog No. 5000223451
Encompass_Preferred
Estramustine phosphate sodium is an estradiol analog and an orally active antimicrotubule chemotherapy research reagent. It depolymerizes microtubules by binding tubulin and microtubule-associated proteins, interfering with mitosis and inducing apoptosis, and is commonly used in cellular and cancer research models.
  • Depolymerizes microtubules and disrupts mitosis.
  • Induces apoptosis, useful for cell-death studies.
  • Suitable for prostate cancer and cell-cycle research models.
  • Provided as the water-soluble disodium phosphate form for handling in aqueous systems.
  • Stable as a solid at 4°C; in solvent store at -80°C or -20°C depending on duration.
Catalog No. 50-002-23451 Supplier Medchemexpress LLC Supplier No. HY136275MG
Estramustine phosphate sodium is an estradiol analog and an orally active antimicrotubule chemotherapy research reagent. It depolymerizes microtubules by binding tubulin and microtubule-associated proteins, interfering with mitosis and inducing apoptosis, and is commonly used in cellular and cancer research models.
  • Depolymerizes microtubules and disrupts mitosis.
  • Induces apoptosis, useful for cell-death studies.
  • Suitable for prostate cancer and cell-cycle research models.
  • Provided as the water-soluble disodium phosphate form for handling in aqueous systems.
  • Stable as a solid at 4°C; in solvent store at -80°C or -20°C depending on duration.