Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estriol 3-Sulfate Sodium Salt, Sigma-Aldrich

Catalog No. 01258929 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-929 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-929 Supplier MilliporeSigma Supelco Supplier No. E637525MG
Add to Cart
Add to Cart

Package size based on steroid content.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Weight (g/mol) 390.43 g/mol

Specifications

Quantity 25 mg
Percent Purity ≥98%
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.