Learn More
Medchemexpress LLC Estriol | 50-27-1 | 500 MG
SDP
Catalog No. 501639404
Encompass_Preferred
Estriol (Oestriol) is an orally active estrogen and an ERα and ERβ agonist. It is also a potent GPR30 antagonist in estrogen receptor-negative breast cancer cells. Estriol can ameliorate disease severity through immunomodulatory mechanisms that decrease tissue inflammation and has powerful proconvulsant effects.
  • Orally active estrogen
  • ERα and ERβ agonist
  • Potent GPR30 antagonist in estrogen receptor-negative breast cancer cells
  • Ameliorates disease severity through immunomodulatory mechanisms that decrease tissue inflammation
  • Has powerful proconvulsant effects
Catalog No. 50-163-9404 Supplier Medchemexpress LLC Supplier No. HYB0412500MG
Estriol (Oestriol) is an orally active estrogen and an ERα and ERβ agonist. It is also a potent GPR30 antagonist in estrogen receptor-negative breast cancer cells. Estriol can ameliorate disease severity through immunomodulatory mechanisms that decrease tissue inflammation and has powerful proconvulsant effects.
  • Orally active estrogen
  • ERα and ERβ agonist
  • Potent GPR30 antagonist in estrogen receptor-negative breast cancer cells
  • Ameliorates disease severity through immunomodulatory mechanisms that decrease tissue inflammation
  • Has powerful proconvulsant effects