Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estriol-d2 | 53866-32-3 | 98.5% | 1 MG
SDP

Catalog No. 5000407876
Encompass_Preferred

Estriol-d2 is the deuterium labeled form of Estriol. It acts as an antagonist of the G-protein coupled estrogen receptor in estrogen receptor-negative breast cancer cells. This compound is suitable for use as a tracer and as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.

  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Deuterium labeled Estriol
  • Antagonist of G-protein coupled estrogen receptor
  • Molecular formula: C18H22D2O3
  • Molecular weight: 290.39
  • Purity: 98.47%
  • Appearance: Solid, white to off-white

Catalog No. 50-004-07876 Supplier Medchemexpress LLC Supplier No. HYB0412S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Estriol-d2 is the deuterium labeled form of Estriol. It acts as an antagonist of the G-protein coupled estrogen receptor in estrogen receptor-negative breast cancer cells. This compound is suitable for use as a tracer and as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.

  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Deuterium labeled Estriol
  • Antagonist of G-protein coupled estrogen receptor
  • Molecular formula: C18H22D2O3
  • Molecular weight: 290.39
  • Purity: 98.47%
  • Appearance: Solid, white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.