Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estriol-d2 | 53866-32-3 | 98.5% | 500 UG
SDP

Supplier:  Medchemexpress LLC HYB0412S500UG

Encompass_Preferred

Estriol-d2 is a deuterium-labeled version of Estriol. It acts as an antagonist of the G-protein coupled estrogen receptor in estrogen receptor-negative breast cancer cells. Intended for research use only, this compound serves as a tracer and internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Deuteration in drug molecules is utilized for quantitation during development and can impact pharmacokinetic and metabolic profiles.

  • Functions as an antagonist of the G-protein coupled estrogen receptor
  • Used as a tracer in research
  • Serves as an internal standard for quantitative analysis
  • Molecular formula: C18H22D2O3
  • Molecular weight: 290.39
  • Available in powder form

Catalog No. 50-004-07788


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.