Learn More
Medchemexpress LLC Estriol-d3 | 79037-36-8 | 97.6% | 1 MG
SDP
Catalog No. 5000409059
Encompass_Preferred
Estriol-d3 is the deuterium labeled Estriol, used as a tracer and an internal standard for quantitative analysis. It acts as an antagonist of the G-protein coupled estrogen receptor in estrogen receptor-negative breast cancer cells. Stable heavy isotopes like Estriol-d3 are incorporated into drug molecules for quantitation during drug development and can influence pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Antagonist of the G-protein coupled estrogen receptor in estrogen receptor-negative breast cancer cells
Catalog No. 50-004-09059 Supplier Medchemexpress LLC Supplier No. HYB0412S21MG
Estriol-d3 is the deuterium labeled Estriol, used as a tracer and an internal standard for quantitative analysis. It acts as an antagonist of the G-protein coupled estrogen receptor in estrogen receptor-negative breast cancer cells. Stable heavy isotopes like Estriol-d3 are incorporated into drug molecules for quantitation during drug development and can influence pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Antagonist of the G-protein coupled estrogen receptor in estrogen receptor-negative breast cancer cells