Learn More
Biomatik Corporation Estriol (E3) CLIA Kit
Supplier:  Biomatik Corporation EKU08622
Encompass_Preferred
Estriol (E3) CLIA Kit for detecting E3 in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 781-200,000pg/ml. Sensitivity: 262.3pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Target Synonyms: oestriol. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7649