Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Estriol ELISA Kit

Catalog No. EIAESTX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIAESTX10 10 x 96 Tests
EIAEST 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EIAESTX10 Supplier Invitrogen™ Supplier No. EIAESTX10
Add to Cart
Add to Cart
Request a Quote

ELISA

The Estriol Competitive ELISA quantitates estriol in serum, plasma, urine, saliva, dried fecal extracts or or cell culture medium.∣Principle of the method: The Estriol Competitive ELISA research-use-only kit is designed to quantitatively measure estriol independent of species. An estriol standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with anti-rabbit IgG antibodies. Anti-rabbit estriol antibodies and estriol-peroxidase conjugate is added to the wells. The estriol-peroxidase conjugate and any estriol in the sample will compete to bind to the anti-rabbit estriol antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound estriol-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of estriol present in the sample.∣Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Estriol is the main estrogen in the blood and urine of pregnant females. Serum estriol determinations reflect the status of the fetus and are used to evaluate fetal distress and placental function.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number C18H24O3; Estratriol; Estriol; Oestriol; Ovestin; Ovestrion
Assay Range 19.66-12000 pg/mL
Assay Sensitivity 5.33 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine
Sufficient For 10 x 96 Assays
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias C18H24O3; Estratriol; Estriol; Oestriol; Ovestin; OvestrionC18H24O3; Estratriol; Estriol; Oestriol; Ovestin; Ovestrion;
Interassay CV 12%
Intraassay CV 10%
Kit Contents Anti-IgG coated 96 well plate, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Fecal Extract 50 μL, Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 12.5 μL
Shipping Condition Wet or Dry Ice
Storage Requirements -20°C
Target Estriol
Test Time 45 min
Total Assay Time 2 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.