Learn More
Cayman Chemical EstrIol ELISA Stndard 1ea
SDP
Catalog No. 502896793
Encompass_Preferred
An inhibitor of DCLK1 (IC50 17 nM) selective for DCLK1 over LRRK2 BRD4 and ERK5 (IC50s 7000 >10000 and 3350 nM respectively) reduces the number of cells in primary patient-derived pancreatic cancer organoids at 1 UM reduces plaque area and collagen levels in the aortic arches and serum levels of Tnf-a and Il-6 an ApoE-/- mouse model of high-fat diet-induced atherosclerosis at 10 mg/kg every other day
Catalog No. 50-289-6793 Supplier Cayman Chemical Supplier No. 4822841 EA
An inhibitor of DCLK1 (IC50 17 nM) selective for DCLK1 over LRRK2 BRD4 and ERK5 (IC50s 7000 >10000 and 3350 nM respectively) reduces the number of cells in primary patient-derived pancreatic cancer organoids at 1 UM reduces plaque area and collagen levels in the aortic arches and serum levels of Tnf-a and Il-6 an ApoE-/- mouse model of high-fat diet-induced atherosclerosis at 10 mg/kg every other day