Learn More
Description
Estriol USP is a white to practically white, odorless crystalline powder. It is insoluble in water, is sparingly soluble in alcohol, and is soluble in vegetable oils. Micronized Estriol USP has over 90% of particles that are less than 10 μm in size.
Alternative name(s): Oestriol
Specifications
Specifications
| Chemical Name or Material | Estriol USP (Micronized) |
| Packaging | Bottle, Amber Glass |
| CAS | 50-27-1 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 288.38 |
| Quantity | 1 g |
| Percent Purity | 97 to 102 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
