Learn More
Biogenex Laboratories Estrogen : PS2.1 : 6mL
SDP
Supplier:  Biogenex Laboratories AM1905M
Encompass_Preferred
This antibody specifically recognizes 6.5 kD human pS2 Estrogen-Regulated Protein. pS2 is specifically expressed and secreted by ER-mucosa cells of the normal stomach (antrum and body) of both female and male individuals. Primary breast tumors have been shown to express pS2 in ER+ primary breast tumors.
Catalog No. 50-197-8753