Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Estrogen Receptor Alpha Activation Antibody Sampler Kit, Unconjugated, 1 Kit
SDP

Catalog No. 502051387
Encompass

The Estrogen Receptor α Activation Antibody Sampler Kit provides an economical solution for detecting the activation of estrogen receptor α using a combination of phospho-specific and control antibodies. This kit is designed for researchers investigating ERα phosphorylation at critical regulatory sites.

  • Includes antibodies for detection of ERα and phosphorylation at Ser118 and Ser167.
  • Supplied with HRP-linked anti-rabbit and anti-mouse secondary antibodies for western blotting.
  • Contains sufficient reagents to perform two western blot experiments per primary antibody.
  • Formulated in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/ml BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Storage requires maintenance at –20°C without aliquoting.

Catalog No. 50-205-1387 Supplier Cell Signaling Technology Supplier No. 49757T
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Estrogen Receptor α Activation Antibody Sampler Kit provides an economical solution for detecting the activation of estrogen receptor α using a combination of phospho-specific and control antibodies. This kit is designed for researchers investigating ERα phosphorylation at critical regulatory sites.

  • Includes antibodies for detection of ERα and phosphorylation at Ser118 and Ser167.
  • Supplied with HRP-linked anti-rabbit and anti-mouse secondary antibodies for western blotting.
  • Contains sufficient reagents to perform two western blot experiments per primary antibody.
  • Formulated in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/ml BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Storage requires maintenance at –20°C without aliquoting.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.