Learn More
Medchemexpress LLC Estrogen receptor modulator 1 | 63676-22-2 | 99.9% | 242.29 | 25 MG
SDP
Catalog No. 5000345627
Encompass_Preferred
Estrogen receptor modulator 1 (compound 18) is an orally active and selective estrogen receptor modulator (SERM), with a pIC50 of 0.46. It induces regression of Tamoxifen-resistant, hormone-independent xenograft tumors.
  • Activation of estrogen response element in HeLa cells stably transfected with human Estrogen receptor beta (EC50: 17 nM).
  • Activation of estrogen response element in HeLa cells stably transfected with human Estrogen receptor alpha (EC50: 50 nM).
  • Agonist activity in transcriptional activation assay in SH-SY5Y neuroblastoma cells expressing Estrogen receptor beta (EC50: 10 nM).
  • Inhibits colony formation at 100 nM for 10 days.
  • Significantly inhibits growth of cells and induces apoptosis at 100 nM for 9 days.
  • Induces regression of T47D:A18/PKCα tumors in vivo at 1.5 mg/animal daily for 2 weeks.
Catalog No. 50-003-45627 Supplier Medchemexpress LLC Supplier No. 25MGHY110201
Estrogen receptor modulator 1 (compound 18) is an orally active and selective estrogen receptor modulator (SERM), with a pIC50 of 0.46. It induces regression of Tamoxifen-resistant, hormone-independent xenograft tumors.
  • Activation of estrogen response element in HeLa cells stably transfected with human Estrogen receptor beta (EC50: 17 nM).
  • Activation of estrogen response element in HeLa cells stably transfected with human Estrogen receptor alpha (EC50: 50 nM).
  • Agonist activity in transcriptional activation assay in SH-SY5Y neuroblastoma cells expressing Estrogen receptor beta (EC50: 10 nM).
  • Inhibits colony formation at 100 nM for 10 days.
  • Significantly inhibits growth of cells and induces apoptosis at 100 nM for 9 days.
  • Induces regression of T47D:A18/PKCα tumors in vivo at 1.5 mg/animal daily for 2 weeks.