Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estrogen receptor modulator 1 | 63676-22-2 | 99.91% | 242.29 | 500 MG
SDP

Catalog No. 5000344012
Encompass_Preferred

Estrogen receptor modulator 1 (compound 18) is an orally active and selective estrogen receptor modulator (SERM), with a pIC50 of 0.46. It induces regression of Tamoxifen-resistant, hormone independent xenograft tumors.

  • Orally active and selective estrogen receptor modulator (SERM).
  • Induces regression of Tamoxifen-resistant, hormone independent xenograft tumors.
  • Inhibits T47D:A18/PKCα and T47D:A18-TAM1 colony formation at 100 nM for 10 days.
  • Significantly inhibits the growth of MCF-7:5C cells and induces apoptosis at 100 nM for 9 days.
  • Appearance: Solid, White to off-white.
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: Powder at -20°C for 3 years; In solvent at -80°C for 6 months, or -20°C for 1 month.

Catalog No. 50-003-44012 Supplier Medchemexpress LLC Supplier No. HY11020110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Estrogen receptor modulator 1 (compound 18) is an orally active and selective estrogen receptor modulator (SERM), with a pIC50 of 0.46. It induces regression of Tamoxifen-resistant, hormone independent xenograft tumors.

  • Orally active and selective estrogen receptor modulator (SERM).
  • Induces regression of Tamoxifen-resistant, hormone independent xenograft tumors.
  • Inhibits T47D:A18/PKCα and T47D:A18-TAM1 colony formation at 100 nM for 10 days.
  • Significantly inhibits the growth of MCF-7:5C cells and induces apoptosis at 100 nM for 9 days.
  • Appearance: Solid, White to off-white.
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: Powder at -20°C for 3 years; In solvent at -80°C for 6 months, or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.