Learn More
Cayman Chemical Estron 3-methyl ether 1g
SDP
Catalog No. 502940771
Encompass_Preferred
A synthetic estrogen; stimulates proliferation of estrogen-dependent MCF-7 breast cancer cells in vitro from 0.16-20 μM; lacks ER binding activity (IC50s = >100 μM for ERα and ERβ); has been used in the synthesis of various estrogen receptor modulators
Catalog No. 50-294-0771 Supplier Cayman Chemical Supplier No. 279781 G
Add to Cart
Add to Cart
A synthetic estrogen; stimulates proliferation of estrogen-dependent MCF-7 breast cancer cells in vitro from 0.16-20 μM; lacks ER binding activity (IC50s = >100 μM for ERα and ERβ); has been used in the synthesis of various estrogen receptor modulators